missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Purified Suitable for Histology, Reagents

Catalog No. 01337620
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
Packaging:
Glass Bottle
HDPE
HDPE Winpak
Poly Bottle
Steel Drum
Steel Pail
Tin Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337620 10 L Steel Pail
01337619 1 L Glass Bottle
01337618 200 L Steel Drum
01337617 20 L HDPE Winpak
01337616 20 L Steel Pail
01337615 20 L HDPE
01337614 4 L Poly Bottle
01337613 4 L Tin Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01337620 Supplier Labchem Inc Supplier No. C210250010E6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetone, Purified Suitable for Histology, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Quantity 10 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.788
Boiling Point 56°C
Grade Pure
Chemical Name or Material Acetone, Purified Suitable for Histology