missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone oxime, 98%
Description
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
Solubility
Soluble in water (330 g/L at 20°C).
Notes
Store away from oxidizing agents. Protect against electrostatic charges.
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
Solubility
Soluble in water (330 g/L at 20°C).
Notes
Store away from oxidizing agents. Protect against electrostatic charges.
Specifications
Specifications
| Melting Point | 60°C to 63°C |
| Density | 0.901 |
| Boiling Point | 133°C to 135°C |
| Flash Point | 60°C (140°F) |
| Quantity | 50 g |
| UN Number | UN1325 |
| Beilstein | 1560146 |
| Merck Index | 14,75 |
| Solubility Information | Soluble in water (330g/L at 20°C). |
| Formula Weight | 73.1 |
| Show More |
Safety and Handling
| Hazard Category | H228-H318-H373 |
| Hazard Statement | GHS H Statement H228 Flammable solid. |
| Precautionary Statement | P210-P240-P241-P260-P280-P305+P351+P338-P310-P314-P370+P378q-P501c |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| EINECSNumber | 204-820-1 |
| RTECSNumber | AL6825000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only