accessibility menu, dialog, popup

UserName

CAS RN 127-06-0

IUPAC Name: N-(propan-2-ylidene)hydroxylamine
Synonyms: acetoxime|ccris 5|actone oxime|acetoxime, 2-(14)C-labeled|acetoxime, 2-(14)c-labeled|2-(14)C-labeled acetoxime|2-(14)c-labeled acetoxime
Molecular Weight (g/mol): 73.1
Molecular Formula: C3H7NO
InChi Key: PXAJQJMDEXJWFB-UHFFFAOYSA-N
SMILES: CC(C)=NO
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)