missing translation for 'onlineSavingsMsg'
Learn More

Acetone oxime, 98%

Catalog No. AAA1080218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1080218 50 g
AAA1080230 250 g
AAA108020B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1080218 Supplier Thermo Scientific Chemicals Supplier No. A1080218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 127-06-0 | C3H7NO | 73.10 g/mol

It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

Solubility
Soluble in water (330 g/L at 20°C).

Notes
Store away from oxidizing agents. Protect against electrostatic charges.

Chemical Identifiers

CAS 127-06-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00002118
InChI Key PXAJQJMDEXJWFB-UHFFFAOYSA-N
Synonym acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
PubChem CID 67180
ChEBI CHEBI:15349
IUPAC Name N-propan-2-ylidenehydroxylamine
SMILES CC(C)=NO

Specifications

Melting Point 60°C to 63°C
Density 0.901
Boiling Point 133°C to 135°C
Flash Point 60°C (140°F)
Quantity 50 g
UN Number UN1325
Beilstein 1560146
Merck Index 14,75
Solubility Information Soluble in water (330g/L at 20°C).
Formula Weight 73.1
Percent Purity 98%
Chemical Name or Material Acetone oxime
Show More Show Less
Hazard Category H228-H318-H373
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P280-P305+P351+P338-P310-P314-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 204-820-1
RTECSNumber AL6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only