missing translation for 'onlineSavingsMsg'
Learn More

Acetone (NF/FCC), Fisher Chemical™

Catalog No. A11S4
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Steel Drum
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A11S4 4 L Aluminum Can
A111 1 L Glass Bottle
A114 4 L Glass Bottle
A1120 20 L Steel Pail
A11200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A11S4 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. BPA11S4
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 7
Melting Point -95°C
Color Colorless
Alkalinity 10mg/kg max. (FCC)
Decomposition Information >4°C
Density 0.790g/cm³
Quantity 4 L
Flash Point -20°C (-4°C)
Identification Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Packaging Aluminum Can
Physical Form Liquid
Refractive Index 1.358 to 1.360 (at 25°C) (FCC)
Specific Gravity 0.789 Max. (NF), 0.7880 max. (FCC), 0.789 max. (NF)
Vapor Pressure 247mbar at 20°C
Linear Formula CH3COCH3
Viscosity 0.32 mPa/s at 20°C
UN Number 1090; Colorless
Water 0.5% Max. (FCC/NF), 0.5% max. (NF)
Boiling Point 56°C
Formula Weight 58.08
Percent Purity ≥99.0% (NF)
Grade FCC/NF
Chemical Name or Material Acetone
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid