missing translation for 'onlineSavingsMsg'
Learn More

Acetone (NF/FCC/EP), Fisher Chemical™

$480.46 - $480.46

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A94
SDS View Documents
Fisher Chemical
A94
4 L EcoSafPak™ Glass Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O

Specifications

7
67-64-1
10mg/kg max. (FCC)
>4°C
56°C
-20°C (-4°C)
EcoSafPak™ Glass Bottle
1.358 to 1.360 (at 25°C), 1.358 to 1.360 (at 25°C) (FCC)
0.789 Max. (NF), 0.7880 max. (FCC)
CH3COCH3
247mbar at 20°C
1090; Colorless
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(C)=O
58.08
CHEBI:15347
NLT 99.5%, NMT 100.5% (FCC)
Acetone
-95°C
Colorless
Pass Test (EP)
0.790g/cm³
4 L
Pass Test, Pass Test (EP), Pass Test (FCC), Pass Test (NF)
Liquid
50ppm max. (EP)
C3H6O
MFCD00008765
0.32 mPa/s at 20°C
≤3g/L (EP), 0.5% max. (FCC), 0.5% max. (NF), 3g/L max. (EP)
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
BP/EP/FCC/JP/NF/USP
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open f

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'