missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, meets ASTM D 329, Reagents
Specifications
Specifications
| Melting Point | -94.7°C |
| Color | Colorless |
| Boiling Point | 56°C |
| Quantity | 200 L |
| Packaging | Steel Drum |
| Physical Form | Liquid |
| Specific Gravity | 0.788 |
| Certifications/Compliance | ASTM |
| Chemical Name or Material | Acetone |