missing translation for 'onlineSavingsMsg'
Learn More

Acetone, meets ASTM D 329, Reagents

Catalog No. 01338190
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338190 200 L
01338189 20 L
01338188 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338190 Supplier Labchem Inc Supplier No. C1102550200L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetone, meets ASTM D 329, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Quantity 200 L
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.788
Certifications/Compliance ASTM
Boiling Point 56°C
Chemical Name or Material Acetone