missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Laboratory Reagent, ≥99.5%, Honeywell™

Catalog No. p-7120103
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045019 1 L Glass Bottle
60045022 4 L Glass Bottle
60045025 6 x 1 L Glass Bottle
60045023 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60045019 Supplier Honeywell Inc Supplier No. 1799731L
Only null left

Laboratory Reagent,≥99.5%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Color Colorless
Density 0.791g/mL at 25°C
Boiling Point 56°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.359(lit.)
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number UN1090
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99.5%
Grade Laboratory Reagent
Chemical Name or Material Acetone
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation class 3 - PG 2 - Acetone
EINECSNumber 200-662-2
RTECSNumber AL3150000