missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, HPLC Grade, ≥ 99.5%, LabChem™
Supplier: LabChem LC104254
Specifications
| Acetone | |
| 67-64-1 | |
| Colorless | |
| ≤0.0003meq/g | |
| Carbon monoxide, carbon dioxide | |
| 56°C | |
| -20°C (-4°C) | |
| Amber Glass | |
| ≤5ppm | |
| CH3COCH3 | |
| 0.417mm2/s | |
| ≤0.5% | |
| Soluble in water | |
| CC(C)=O | |
| 58.08 | |
| CHEBI:15347 | |
| ≥99.5% |
| -95°C | |
| Passes Test | |
| 100 | |
| ≤0.0006meq/g | |
| 0.79kg/m3 | |
| 4 L | |
| Passes Test | |
| Liquid | |
| C3H6O | |
| MFCD00008765 | |
| 1090; Colorless | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 180 | |
| 58.08 | |
| HPLC |
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| 180 | |
| propan-2-one |
| C3H6O | |
| MFCD00008765 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CHEBI:15347 | |
| CC(C)=O |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/hot surfaces, open flames.
No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only
Danger
DOTInformation : Class 3: Flammable Liquid
Recommended Storage : Room Temperature