missing translation for 'onlineSavingsMsg'
Learn More

Acetone (HPLC), Fisher Chemical™

Catalog No. p-215618
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
EcoSafPak™ Amber Glass Bottle
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A949SK1 1 L Safe-Cote™ Amber Glass 
A9491 1 L Wide Neck Amber Bottle
A9494 4 L EcoSafPak™ Amber Glass Bottle
A949SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A949SK1 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. BPA949SK 1
Only null left

HPLC mobile phase for analytical and preparative separations

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 7
Melting Point -95°C
Absorbance 0.01 AU max. at 400nm, 0.02 AU max. at 350nm, 0.06 AU max. at 340nm, 1.00 AU max. at 330nm
Color Colorless
Decomposition Information >4°C
Density 0.790g/cm³
Quantity 1 L
Identification Pass Test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Residue after Evaporation 1ppm max.
Assay Percent Range 99.5% min by GC
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0006mEq/g max.
Linear Formula CH3COCH3
Vapor Pressure 247mbar at 20°C
Viscosity 0.32 mPa/s at 20°C
Water 0.5% max.
Boiling Point 56°C
Formula Weight 58.08
Percent Purity ≥99.5%
Grade Mobile phase for HPLC applications
Chemical Name or Material Acetone
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid