missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Histology grade, 99% min, MilliporeSigma™

Catalog No. MAX012520
Change view
Click to view available options
Quantity:
200 L
Packaging:
Metal Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX012520 200 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX012520 Supplier Technidata Supplier No. AX012520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Color Colorless
Quantity 200 L
Identification To pass test
Infrared Spectrum Conforms to standard
Packaging Metal Drum
Physical Form Liquid
Residue after Evaporation 0.001% max
Assay Percent Range 99% min.
Formula Weight 58.08
Percent Purity ≥99% (GC)
Chemical Name or Material Acetone, For analysis Histology
Show More Show Less
Recommended Storage Room Temperature