missing translation for 'onlineSavingsMsg'
Learn More

Acetone, For pesticide residue analysis, Solstice

Catalog No. 60046342
Change view
Click to view available options
Quantity:
1 L
4 x 2.5 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046342 4 x 2.5 L
6000210 1 L
6000212 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046342 Supplier Honeywell Inc Supplier No. 344804X2.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point −17°C
Packaging Glass Bottle
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Boiling Point 55°C to 57°C
Formula Weight 58.08
Percent Purity >99.8%
Grade Pesticide
Chemical Name or Material Acetone, For pesticide residue analysis
Show More Show Less
ShelfLife 720 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone
EINECSNumber 200-662-2
Recommended Storage Room Temp