missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Environmental Grade, 99.5+%

Catalog No. AA43053K2
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43053K2 1 L
AA43053K7 4 L
AA43053M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43053K2 Supplier Thermo Scientific Chemicals Supplier No. 043053K2

Suitable For Pesticide Residue Analysis | CAS: 67-64-1 | C3H6O | 58.08 g/mol

Suitable for pesticide residue analysisAcetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glassware due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon and photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point °C to 94°C
Color Colorless
Density 0.791
Quantity 1 L
Flash Point -17°C (1°F)
Ignition Point 465°C (869°F)
Physical Form Liquid
Refractive Index 1.359
Assay Percent Range >99.5%
Vapor Pressure 185mm Hg at 20°C
UN Number 1090; Colorless
Beilstein 635680
Boiling Point 56°C
Solubility Information Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils.
Formula Weight 58.08
Grade Environmental
Chemical Name or Material Acetone
Show More Show Less
Hazard Category H225-H319-H336-H373
Hazard Statement Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. If swallowed seek medical advice immediately and show this container or label.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 200-662-2
RTECSNumber AL3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only