missing translation for 'onlineSavingsMsg'
Learn More

Acetone, For HPLC, MilliporeSigma™

Catalog No. 50947861
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle (Pre-packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50947861 4 L Glass Bottle (Pre-packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947861 Supplier Technidata Supplier No. AX0115P1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.4°C
Quantity 4 L
Flash Point ≥20°C
Odor Fruity
Packaging Glass Bottle (Pre-packed)
Physical Form Clear liquid, free from particulates
Residue after Evaporation 5ppm
Titratable Acid 0.0003meq/g
Vapor Pressure 233 hPa (20°C)
Water 0.5%
UN Number UN 1090
Boiling Point 56.2°C (1013 hPa)
Formula Weight 58.08
Percent Purity 99.5% min
Chemical Name or Material Acetone
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category H225 Highly flammable liquid and vapor. H319 Causes serious eye irritation. H336 May cause drowsiness or dizziness.
Hazard Statement P210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. P233 Keep container tightly closed. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
EINECSNumber 200-662-2
RTECSNumber AL3150000