missing translation for 'onlineSavingsMsg'
Learn More

Acetone, CHROMASOLV™ Plus, for HPLC, Honeywell™

Catalog No. p-7122141
Change view
Click to view available options
Quantity:
1 L
4 x 4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000208 1 L
6000209 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000208 Supplier Honeywell Inc Supplier No. 6505011L

CHROMASOLV™ Plus, for HPLC,≥99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Linear Formula CH3COCH3
Vapor Pressure 184 mmHg (20°C)
UN Number UN1090
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material Acetone
Show More Show Less
Hazard Statement P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235
RTECSNumber AL3150000