missing translation for 'onlineSavingsMsg'
Learn More

Acetone, B&J Brand™, for HPLC, spectrophotometry (not for pesticide residue analysis), >99.9% (excluding water), Honeywell Burdick & Jackson
GREENER_CHOICE

Catalog No. 6000213
Change view
Click to view available options
Quantity:
200 L
Packaging:
Returnable Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000213 200 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000213 Supplier Honeywell Chemicals Supplier No. CS010200
Only null left

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Absorbance 0.06 AU max. at 340 nm, 0.01 AU max. at 350 nm, 0.005 AU max. at 375 nm, 0.005 AU max. at 400 nm
CAS Max % 100.0000%
Density 0.79 g/cm3
Quantity 200 L
Flash Point −20°C
Packaging Returnable Steel Drum
UN Number UN1090
Water 0.5% max.
Beilstein 635680
Boiling Point 56°C
Solubility Information Miscible in Water at 20°C
Formula Weight 58.08
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Acetone, B&J Brand™, for HPLC, spectrophotometry (not for pesticide residue analysis), >99.9% (excluding water)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone