missing translation for 'onlineSavingsMsg'
Learn More

Acetone, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (excluding water), Honeywell Burdick & Jackson

Catalog No. p-7111713
Change view
Click to view available options
Quantity:
1 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000217 6 x 1 L Glass Bottle
6000216 1 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000217 Supplier Honeywell Chemicals Supplier No. 0101L
Only null left

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Absorbance 0.06 AU max. at 340 nm (UV), 0.01 AU max. at 350 nm (UV), 0.005 AU max. at 375 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Density 0.79 g/cm3
Quantity 6 x 1 L
Flash Point −20°C
Packaging Glass Bottle
Water 0.5% max.
UN Number UN1090
Beilstein 635680
Boiling Point 56°C
Solubility Information Miscible in Water at 20°C
Formula Weight 58.08
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Acetone, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (excluding water)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone