missing translation for 'onlineSavingsMsg'
Learn More

Acetone, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (excluding water), Solstice

Catalog No. 6000214
Change view
Click to view available options
Quantity:
20 L
Packaging:
HDPE Nowpak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000214 20 L HDPE Nowpak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000214 Supplier Solstice Supplier No. NP01020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Max % 100.0000%
Density 0.79 g/cm3
Quantity 20 L
Flash Point −20°C
Packaging HDPE Nowpak™
UN Number UN1090
Beilstein 635680
Boiling Point 56°C
Formula Weight 58.08
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Acetone, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (excluding water)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone