missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS Reagent Grade, Ricca Chemical

Catalog No. RSOA00104A
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOA00104A 4 L Natural Poly Bottle
RSOA001020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RSOA00104A Supplier Ricca Chemical Company Supplier No. RSOA00104A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Quantity 4 L
Packaging Natural Poly Bottle
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0003 meq/g
Titratable Base max. 0.0006 meq/g
Water max. 0.5%
Synonym acetone
Boiling Point 56°C
Percent Purity min. 99.5%
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
ShelfLife 60 months