missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS Reagent, Reagents

Catalog No. 01338198
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Bottle
Safety Coated Glass
Steel Drum
Steel Pail
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338198 1 L Glass Bottle
01338199 10 L Steel Pail
01338197 200 L Steel Drum
01338196 20 L Steel Pail
01338195 20 L HDPE Winpak
01338194 4 L Poly Bottle
01338193 4 L Glass Bottle
01338192 4 L Safety Coated Glass
01338191 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 01338198 Supplier Labchem Inc Supplier No. C11025001C
Only null left

Acetone, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.788
Boiling Point 56°C
Grade ACS
Chemical Name or Material Acetone, ACS Reagent