missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, ACS Reagent, Reagents
Acetone, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1102500200E
Specifications
| Acetone, ACS Reagent | |
| 67-64-1 | |
| 56°C | |
| Steel Drum | |
| 0.788 | |
| MFCD00008765 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| ACS |
| -94.7°C | |
| Colorless | |
| 200 L | |
| Liquid | |
| C3H6O | |
| Dimethyl Ketone, 2-Propanone | |
| CC(C)=O | |
| 58.08 |
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one |
| C3H6O | |
| MFCD00008765 | |
| Dimethyl Ketone, 2-Propanone | |
| CC(C)=O |