missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, ACS Reagent, Reagents
Specifications
Specifications
| Melting Point | -94.7°C |
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Safety Coated Glass |
| Physical Form | Liquid |
| Specific Gravity | 0.788 |
| Boiling Point | 56°C |
| Grade | ACS |
| Chemical Name or Material | Acetone, ACS Reagent |