missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Solstice

Catalog No. 6000224
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000224 4 x 4 L Glass Bottle
6000225 4 x 4 L Poly Bottle
6000222 19 L Metal Can
6000223 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000224 Supplier Solstice Supplier No. LP0104

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Max % 100.0000%
Density 0.79 g/cm3
Quantity 4 x 4 L
Flash Point −20°C
Packaging Glass Bottle
Water 0.5% max.
UN Number UN1090
Beilstein 635680
Boiling Point 56°C
Solubility Information Miscible in Water at 20°C
Formula Weight 58.08
Percent Purity >99.5%
Grade ACS
Chemical Name or Material Acetone, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone