missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell
Supplier: Honeywell Chemicals LP01019
Specifications
| Acetone | |
| 67-64-1 | |
| 0.79 g/cm3 | |
| 19 L | |
| Metal Can | |
| MFCD00008765 | |
| 0.5% max. | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 180 | |
| 58.08 | |
| ACS Reagent |
| ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5% | |
| 100.0000% | |
| 56°C | |
| −20°C | |
| C3H6O | |
| UN1090 | |
| 635680 | |
| Miscible in Water at 20°C | |
| CC(C)=O | |
| 58.08 | |
| CHEBI:15347 | |
| >99.5% |
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| 180 | |
| propan-2-one |
| C3H6O | |
| MFCD00008765 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CHEBI:15347 | |
| CC(C)=O |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone