missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™
ACS reagent,≥99.5%
Supplier: Honeywell-Riedel-de Haen 1791244X25L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Acetone | |
| ACS reagent, ≥99.5% | |
| ≥90.0000% | |
| 0.791Â g/mLÂ at 25Â °C(lit.) | |
| 4 x 2.5 L | |
| Glass Bottle | |
| C3H6O | |
| MFCD00008765 | |
| 635680 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 180 | |
| 58.08 | |
| ACS Reagent |
| -94°C (literature) | |
| 67-64-1 | |
| ≤100.0000% | |
| 55°C to 57°C | |
| −17°C | |
| acc. to ACS | |
| CH3COCH3 | |
| UN1090 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CC(C)=O | |
| 58.08 | |
| CHEBI:15347 | |
| >99.5% |
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| 180 | |
| propan-2-one |
| C3H6O | |
| MFCD00008765 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CHEBI:15347 | |
| CC(C)=O |
Safety and Handling
ShelfLife : 1080 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone
EINECSNumber : 200-662-2
Recommended Storage : Room Temp