missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS Grade, ≥ 99.5%, LabChem™

Catalog No. p-5379502
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 4 L
Packaging:
Amber Glass
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC104201 500 mL Amber Glass
LC104202 1 L Amber Glass Bottle
LC104204 4 L Amber Glass
LC1042044 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC104201 Supplier LabChem Supplier No. LC104201
Only null left

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Color Colorless
CAS Max % 100
Acidity ≤0.0003meq/gcSt
Alkalinity ≤0.0006meq/gcSt
Decomposition Information Carbon monoxide, Carbon dioxide
Density 786kg/m3
Quantity 500 mL
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤0.001%
Linear Formula CH3COCH3
Viscosity 0.417mm2/s
Water ≤0.5%
UN Number UN1090
Solubility Information Soluble in water
Formula Weight 58.08
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness and dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/hot surfaces, open flames.
No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist/vapors/spray.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
If inhaled: Remove person to fresh air and keep at rest in a posittion comfortable for breathing.
Call a poison center/doctor if you feel unwell.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container in accordance with local, state and federal regulations.
DOTInformation DOT Class 3: Flammable Liquid
Recommended Storage Room Temperature