missing translation for 'onlineSavingsMsg'
Learn More

Acetone, 99+%, for spectroscopy

Catalog No. AC167645000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167645000 500 mL Glass Bottle
AC167640025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167645000 Supplier Thermo Fisher Scientific Formerly Acros Organics Supplier No. 167645000

CAS: 67-64-1 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Absorbance 0.01 max. at 400nm, 0.02 max. at 350nm, 0.1 max. at 340nm, 1 max. at 330nm
Decomposition Information >4°C
Density 0.7900g/mL
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range 99+%
Vapor Pressure Vapor Pressure: 247mbar at 20°C
Linear Formula CH3COCH3
Water 0.5% max. (Coulometric)
Beilstein 01, 635
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Formula Weight 58.08
Percent Purity ≥99.0%
Grade Spectroscopy
Chemical Name or Material Acetone, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-662-2
RTECSNumber AL3150000
TSCA TSCA

RUO – Research Use Only