missing translation for 'onlineSavingsMsg'
Learn More

Acetone, 99%, MilliporeSigma™

Catalog No. 50947612
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947612 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947612 Supplier Technidata Supplier No. AX01253

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 5 to 6 (395 g/L, H2O, 20°C)
Melting Point −95.4°C
Quantity 20 L
Flash Point < −20°C
Packaging Steel Drum
Residue after Evaporation 0.001% max
Assay Percent Range 99% min
Vapor Pressure 233 hPa (20°C)
Boiling Point 56.2°C (1013 hPa)
Recommended Storage Store at room temperature