missing translation for 'onlineSavingsMsg'
Learn More

Acetone, 99.8%, for HPLC
GREENER_CHOICE

Catalog No. AC268310010
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC268310010 1 L
AC268310025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268310010 Supplier Thermo Scientific Chemicals Supplier No. 268310010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-64-1 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
Absorbance (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 350nm, 0.005 max. at 360nm, 0.005 max. at 370nm, 0.09 max. at 340nm, 0.3 max. at 335nm, 1 max. at 330nm
CAS Min % 99.75
Acidity ≤0.0003meq/g
CAS Max % 100.0
Alkalinity ≤0.0003meq/g
Decomposition Information >4°C
Density 0.7900g/mL
Quantity 1 L
Flash Point −20°C
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3585
Residue after Evaporation ≤0.0002%
Specific Gravity 0.79
Assay Percent Range 99.8%
Linear Formula CH3COCH3
Vapor Pressure Vapor Pressure: 247mbar at 20°C
Viscosity 0.32 mPa.s (20°C)
UN Number 1090; Colorless
Water ≤0.1%
Beilstein 01, IV, 3180
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Green Features ACT Ecolabel (Gold)
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Boiling Point 56.0°C
Formula Weight 58.08
Percent Purity 99.8%
Grade HPLC
Chemical Name or Material Acetone
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-662-2
RTECSNumber AL3150000
TSCA TSCA

RUO – Research Use Only