missing translation for 'onlineSavingsMsg'
Learn More

Acetone, 99.5% min, ACS, MilliporeSigma™

Catalog No. MAX0120P4
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Metal Can
Poly Bottle
Poly Drum
Poly-coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX0120P4 4 L Poly-coated Glass Bottle
MAX01206 500 mL Glass Bottle
MAX01209 1 L Poly Bottle
MAX01208 4 L Glass Bottle
MAX01203 20 L Metal Can
MAX012030 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MAX0120P4 Supplier Technidata Supplier No. US1AX0120P/4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.2°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 4 L
Flash Point -18.15°C (-0.67°F) Closed cup
Ignition Point 464.85°C (868.7°F)
Odor Pungent
Packaging Poly-coated Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max
Assay Percent Range 99.5% min.
Titratable Acid 0.0003meq/g max
Titratable Base 0.0006meq/g max
Linear Formula CH3COCH3
Water 0.5% max
UN Number UN 1090
Boiling Point 56.1°C
Formula Weight 58.08
Percent Purity ≥99.5% (GC, corrected for water content)
Grade ACS/NF/USP
Chemical Name or Material Acetone, For ACS analysis
Show More Show Less
DOTInformation DOT Class 3
Recommended Storage Room Temperature