missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, ≥99.5% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 67-64-1 |
| Vapor Pressure | 184 mmHg (20°C) |
| Vapor Density | 2 (vs. air) |
| Density | 0.791 g/mL (at 25°C) |
| Flash Point | -17°C |
| Physical Form | Neat |
| Linear Formula | CH3 COCH3 |
| MDL Number | MFCD00008765 |
| Melting Point | -94°C |
| Boiling Point | 56°C (760 mmHg) |
| Show More |