missing translation for 'onlineSavingsMsg'
Learn More

Acetone-1,3-dicarboxylic acid, 97%

CAS: 542-05-2 | C5H6O5 | 146.098 g/mol

$90.58 - $1042.32

Chemical Identifiers

CAS 542-05-2
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.098
MDL Number MFCD00002711
InChI Key OXTNCQMOKLOUAM-UHFFFAOYSA-N
Synonym 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure
PubChem CID 68328
IUPAC Name 3-oxopentanedioic acid
SMILES C(C(=O)CC(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1374218
View Documents
Thermo Scientific Chemicals
A1374218
50 g
Each for $90.58
Only null left
 
AAA1374230
View Documents
Thermo Scientific Chemicals
A1374230
250 g
Each for $326.16
Only null left
 
AAA137420B
View Documents
Thermo Scientific Chemicals
A137420B
1000 g
Each for $1,042.32
Only null left
 
Description

Description

Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

Solubility
Soluble in water and alcohol. Slightly soluble in ethyl acetate and ether. Insoluble in chloroform.

Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
Specifications

Chemical Identifiers

542-05-2
146.098
OXTNCQMOKLOUAM-UHFFFAOYSA-N
68328
C(C(=O)CC(=O)O)C(=O)O
C5H6O5
MFCD00002711
1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure
3-oxopentanedioic acid

Specifications

542-05-2
C5H6O5
50 g
Hygroscopic
1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure
OXTNCQMOKLOUAM-UHFFFAOYSA-N
3-oxopentanedioic acid
68328
97%
∼132°C (decomposition)
MFCD00002711
1447081
14,68
Soluble in water and alcohol. Slightly soluble in ethyl acetate and ether. Insoluble in chloroform.
C(C(=O)CC(=O)O)C(=O)O
146.098
146.1
Acetone-1,3-dicarboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H500

EINECSNumber : 208-797-9

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only