accessibility menu, dialog, popup

UserName

CAS RN 542-05-2

IUPAC Name: 3-oxopentanedioate
Synonyms: acetonedicarboxylic acid
Molecular Weight (g/mol): 144.08
Molecular Formula: C5H4O5
InChi Key: OXTNCQMOKLOUAM-UHFFFAOYSA-L
SMILES: [O-]C(=O)CC(=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)