missing translation for 'onlineSavingsMsg'
Learn More

Acetohydrazide 98.0+%, TCI America™

Catalog No. p-7117056
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A078925G 25 g
A0789100G 100 g
A0789500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A078925G Supplier TCI America Supplier No. A078925G

Chemical Identifiers

CAS 1068-57-1
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.083
MDL Number MFCD00007610
InChI Key OFLXLNCGODUUOT-UHFFFAOYSA-N
Synonym acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
PubChem CID 14039
ChEBI CHEBI:48978
IUPAC Name acetohydrazide
SMILES CC(=O)NN

Specifications

Melting Point 66°C
Color White
Boiling Point 134°C
Quantity 25 g
Formula Weight 74.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acetohydrazide
RTECSNumber AI1225000
TSCA Yes