accessibility menu, dialog, popup

UserName

CH3ONHH2N

CAS RN 1068-57-1

IUPAC Name: acetohydrazide
Synonyms: acetohydrazide hydrazide acetic acid acetyl hydrazide hydrazid kyseliny octove
Molecular Weight (g/mol): 74.08
Molecular Formula: C2H6N2O
InChi Key: OFLXLNCGODUUOT-UHFFFAOYSA-N
SMILES: CC(=O)NN
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)