missing translation for 'onlineSavingsMsg'
Learn More

Acetobromo-alpha-D-glucose, 97%, stab. with 2% calcium carbonate

CAS: 572-09-8 | C14H19BrO9 | 411.20 g/mol

$217.14 - $653.16

Chemical Identifiers

CAS 572-09-8
Molecular Formula C14H19BrO9
Molecular Weight (g/mol) 411.20
MDL Number MFCD00063254
InChI Key CYAYKKUWALRRPA-HAGGZJDPNA-N
Synonym 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromglucose, acetobromoglucose, a-acetobromglucose, acetobromo glucose, acetobromo glucose, a-d-glucopyranosyl bromide, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate
PubChem CID 101776
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0415109
View Documents
Thermo Scientific Chemicals
L0415109
10 g
Each for $217.14
Only null left
 
AAL0415118
View Documents
Thermo Scientific Chemicals
L0415118
50 g
Each for $653.16
Only null left
 
Description

Description

Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.

Solubility
Soluble in ether, chloroform, ethanol, acetone, dichloromethane, ether, ethyl, methanol, and tetrahydrofuran. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
Specifications

Chemical Identifiers

572-09-8
411.20
CYAYKKUWALRRPA-HAGGZJDPNA-N
101776
CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
C14H19BrO9
MFCD00063254
2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromglucose, acetobromoglucose, a-acetobromglucose, acetobromo glucose, acetobromo glucose, a-d-glucopyranosyl bromide, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate
[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate

Specifications

572-09-8
C14H19BrO9
10 g
Moisture sensitive
2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, stab. with calcium carbonate, acetobromoglucose, acetobromo-alpha-d-glucose, a-acetobromglucose, a-acetobromglucose, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate
CYAYKKUWALRRPA-HAGGZJDPNA-N
CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
411.20
411.22
Acetobromo-alpha-D-glucose, stabilized with 2% calcium carbonate
84°C to 89°C
MFCD00063254
96669
14,60
Soluble in ether,chloroform,ethanol,acetone,dichloromethane,ether,ethyl,methanol,and tetrahydrofuran. Insoluble in water.
+198° (c=3 in Chloroform)
[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate
101776
97%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-339-0

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only