Learn More
Acetobromo-alpha-D-glucose, 97%, stab. with 2% calcium carbonate
CAS: 572-09-8 | C14H19BrO9 | 411.20 g/mol
Supplier: Thermo Scientific Chemicals L0415109
Description
Acetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcetobromo-α-D-glucose acts as an intermediate in the preparation of beta-glucosides. It is also used as a possible poly(ethylene terephthalate) surface modification reagent to enhance its blood compatibility.
Solubility
Soluble in ether, chloroform, ethanol, acetone, dichloromethane, ether, ethyl, methanol, and tetrahydrofuran. Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
Specifications
| Acetobromo-alpha-D-glucose | |
| 572-09-8 | |
| C14H19BrO9 | |
| 10 g | |
| Moisture sensitive | |
| 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, stab. with calcium carbonate, acetobromoglucose, acetobromo-alpha-d-glucose, a-acetobromglucose, a-acetobromglucose, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate | |
| CYAYKKUWALRRPA-HAGGZJDPNA-N | |
| CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O | |
| 411.20 | |
| 411.22 |
| stabilized with 2% calcium carbonate | |
| 84°C to 89°C | |
| MFCD00063254 | |
| 96669 | |
| 14,60 | |
| Soluble in ether,chloroform,ethanol,acetone,dichloromethane,ether,ethyl,methanol,and tetrahydrofuran. Insoluble in water. | |
| +198° (c=3 in Chloroform) | |
| [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate | |
| 101776 | |
| 97% |
Chemical Identifiers
| 572-09-8 | |
| 411.20 | |
| CYAYKKUWALRRPA-HAGGZJDPNA-N | |
| 101776 | |
| CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O |
| C14H19BrO9 | |
| MFCD00063254 | |
| 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, stab. with calcium carbonate, acetobromoglucose, acetobromo-alpha-d-glucose, a-acetobromglucose, a-acetobromglucose, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate | |
| [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxan-2-yl]methyl acetate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-339-0
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only