missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoacetanilide, 99%
CAS: 102-01-2 | C10H11NO2 | 177.2 g/mol
Supplier: Thermo Scientific Chemicals 102360010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acetoacetanilide | |
| 102-01-2 | |
| 100.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| CH3COCH2CONHC6H5 | |
| 1 kg | |
| 15, 57 | |
| Solubility in water: 5g/L (20 c). Other solubilities: soluble in acids and alkali hydroxide solutions,soluble in alcohol,chloroform,ether,hot benzene,and hot petroleum ether | |
| CC(=O)CC(=O)NC1=CC=CC=C1 | |
| 177.2 | |
| 177.2 | |
| Crystalline Powder |
| 99% | |
| 98.5 | |
| 83.0°C to 86.0°C | |
| 162°C | |
| 98.5% min. (HPLC) | |
| C10H11NO2 | |
| MFCD00008780 | |
| 12, 518 | |
| acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline | |
| DYRDKSSFIWVSNM-UHFFFAOYSA-N | |
| 3-oxo-N-phenylbutanamide | |
| 7592 | |
| 99% |
Chemical Identifiers
| 102-01-2 | |
| 177.2 | |
| DYRDKSSFIWVSNM-UHFFFAOYSA-N | |
| 7592 | |
| CC(=O)CC(=O)NC1=CC=CC=C1 |
| C10H11NO2 | |
| MFCD00008780 | |
| acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline | |
| 3-oxo-N-phenylbutanamide |
Safety and Handling
EINECSNumber : 202-996-4
RTECSNumber : AK4200000
TSCA : TSCA
RUO – Research Use Only