missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetanilide, 99%

Catalog No. p-48404
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC102360050 5 g
AC102365000 500 g
AC102360010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102360050 Supplier Thermo Scientific Chemicals Supplier No. 102360050
Only null left

CAS: 102-01-2 | C10H11NO2 | 177.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-01-2
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00008780
InChI Key DYRDKSSFIWVSNM-UHFFFAOYSA-N
Synonym acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
PubChem CID 7592
IUPAC Name 3-oxo-N-phenylbutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83.0°C to 86.0°C
Color White
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3COCH2CONHC6H5
Quantity 5 g
Beilstein 12, 518
Merck Index 15, 57
Solubility Information Solubility in water: 5g/L (20 c). Other solubilities: soluble in acids and alkali hydroxide solutions,soluble in alcohol,chloroform,ether,hot benzene,and hot petroleum ether
Formula Weight 177.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetoacetanilide, 99%
Show More Show Less
EINECSNumber 202-996-4
RTECSNumber AK4200000
TSCA TSCA

RUO – Research Use Only