missing translation for 'onlineSavingsMsg'
Learn More

Acetic hydrazide, 96%

CAS: 1068-57-1 | C2H6N2O | 74.083 g/mol

Supplier:  Thermo Scientific Chemicals A1096922

 View more versions of this product

Catalog No. AAA1096922

Only null left
Explore available promotions

Description

Description

Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong bases.
Specifications

Specifications

Acetic hydrazide
∼60°C to 68°C
>110°C (230°F)
MFCD00007610
506165
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
OFLXLNCGODUUOT-UHFFFAOYSA-N
acetohydrazide
14039
74.08
1068-57-1
128°C to 129°C (18 mmHg)
C2H6N2O
100 g
Hygroscopic
Soluble in water.
CC(=O)NN
74.083
CHEBI:48978
96%

Chemical Identifiers

1068-57-1
74.083
OFLXLNCGODUUOT-UHFFFAOYSA-N
14039
acetohydrazide
C2H6N2O
MFCD00007610
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
CHEBI:48978
CC(=O)NN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302+H332-H315-H319-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 213-948-7

RTECSNumber : AI1225000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only