missing translation for 'onlineSavingsMsg'
Learn More

Acetic Anhydride (Certified ACS), Fisher Chemical™

Catalog No. 3510011
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3510011 1 L Glass Bottle
351001100 100 mL Glass Bottle
351001500 500 mL Glass Bottle
3510014 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3510011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Melting Point -73.1°C
Color Colorless
Density 1.087g/cm³
pH 3
Boiling Point 140°C
Flash Point 53°C
Assay Percent Range ≥97.0 %
Packaging Glass Bottle
Linear Formula (CH3CO)2O
Quantity 1 L
UN Number 1715
Residue after Evaporation 0.002% max.
Formula Weight 102.09
Percent Purity ≥97.0%
Grade Certified ACS
Physical Form Liquid
Chemical Name or Material Acetic Anhydride
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive