missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, TraceSELECT™ Ultra, Honeywell Fluka™

Catalog No. p-7071738
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000038 250 mL
6000039 500 mL
6000037 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000038 Supplier Honeywell Chemicals Supplier No. 07692250ML
Only null left

TraceSELECT™ Ultra, for ultratrace analysis,≥99.0%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
Linear Formula CH3CO2H
UN Number 2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity ≥99%
Grade TraceSELECT™ Ultra
Density 1.049g/mL at 25°C
Quantity 250 mL
Explosion Limit 4% (59°F), 16% (92°F)
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Plastic Bottle
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic Acid
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H226-H314
EINECSNumber 200-580-7
RTECSNumber AF1225000