missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, TraceSELECT™, ≥99%, Honeywell Fluka™

TraceSELECT™, for trace analysis,≥99.0%

$371.55 - $371.55

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046914
View Documents
Honeywell Chemicals
45727500ML
500 mL Poly Bottle
Each for $371.55
Only null left
 
Description

Description

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

16.2°C
117°C to 118°C
Liquid
CH3CO2H
UN2789
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
≥99%
1.049g/mL at 25°C
4% at 59°F, 16% at 92°F
800°F
n20/D 1.371
Acetic acid
64-19-7
Colorless
C2H4O2
MFCD00036152
506007
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05g/mol
TraceSELECT
500 mL
40°C (104°F)
Poly Bottle
2.07 (vs air)
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338-P310

DOTInformation : 3(8)/PGII

RTECSNumber : 200-580-7

SDS
Documents

Documents