missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, tested according to Ph.Eur., Honeywell™

Catalog No. 6000036
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000036 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000036 Supplier Honeywell Chemicals Supplier No. 457411L
Only null left

Tested according to Ph.Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05g/mol
Density 1.049g/mL at 25°C
Quantity 1 L
Explosion Limit 4% at 59°F, 16% at 92°F
Flash Point 40°C (104°F)
Ignition Point 800°F
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic acid, tested according to Ph.Eur.
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
RTECSNumber AF1225000