missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid solution, 2.0M, Honeywell™

Catalog No. 6000013
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
6000013 100 mL
1 options

Catalog No. 6000013

Supplier: Honeywell Chemicals A8976100ML

Only null left

Volumetric, 2.0 M CH3COOH (2.0N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Linear Formula CH3CO2H
UN Number UN2790C
Beilstein 506007
Concentration 2.0M
Formula Weight 60.05g/mol
Quantity 100 mL
Chemical Name or Material Acetic acid solution
Hazard Statement P305 + P351 + P338