missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid solution, 0.5M, Honeywell™
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Linear Formula | CH3CO2H |
| UN Number | NONH for all modes of transport |
| Beilstein | 506007 |
| Concentration | 0.5M |
| Formula Weight | 60.05g/mol |
| Quantity | 1 L |
| Chemical Name or Material | Acetic acid solution |