missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, sodium salt, 99+%, for analysis, anhydrous
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Supplier: Thermo Scientific Chemicals 220890010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
| Acetic acid, sodium salt | |
| p.a. | |
| 127-09-3 | |
| 100.0 | |
| 99+% | |
| CH3CO2Na | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium;acetate | |
| 517045 | |
| 82.03 | |
| Analytical | |
| >250°C | |
| 1% max. | |
| Crystals or Crystalline Powder |
| 7.5 to 9.2 (5% soln. at 20°C) | |
| 324.0°C | |
| 99.0 | |
| White | |
| C2H3NaO2 | |
| MFCD00012459 | |
| Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| ≥99% | |
| 1 kg | |
| Authentic | |
| Plastic bottle |
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| [Na+].CC([O-])=O |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 204-823-8
RTECSNumber : AJ4300010
TSCA : TSCA
RUO – Research Use Only