missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt, 99+%, for analysis, anhydrous

Catalog No. p-45214
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220892500 250 g Plastic bottle
AC220890010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC220892500 Supplier Thermo Scientific Chemicals Supplier No. 220892500
Only null left

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

pH 7.5 to 9.2 (5% soln. at 20°C)
Melting Point 324.0°C
CAS Min % 99.0
Color White
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3CO2Na
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name sodium acetate
Formula Weight 82.03
Percent Purity ≥99%
Grade Analytical
Quantity 250 g
Flash Point >250°C
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Plastic bottle
Physical Form Crystals or Crystalline Powder
Chemical Name or Material Acetic acid, sodium salt, p.a.
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 204-823-8
RTECSNumber AJ4300010
TSCA TSCA

RUO – Research Use Only