missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™

Catalog No. 60046219
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
60 kg
6 x 1 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic Pail
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046219 6 x 1 L Glass Bottle
6000007 1 L Glass Bottle
6000006 1 L Plastic Bottle
6000009 2.5 L Glass Bottle
6000008 2.5 L Plastic Bottle
6000011 5 L Plastic Bottle
60046218 6 x 1 L Plastic Bottle
60046217 4 x 2.5 L Glass Bottle
60046216 4 x 2.5 L Plastic Bottle
6000010 30 kg Plastic Pail
6000012 60 kg Plastic Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 60046219 Supplier Honeywell Chemicals Supplier No. 332096X1LGL
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Linear Formula CH3CO2H
UN Number UN2789
Formula Weight 60.05
Percent Purity ≥99.8%
Density 1.049g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 16% at 92°F, 4% at 59°F
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Glass Bottle
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic acid
Show More Show Less
Hazard Statement P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
DOTInformation 3(8) / PGII
RTECSNumber 200-580-7